Learn More
SLC14A1 Antibody (888418), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8238L
Product Code. 18586019
  • € 644.00 / 0.10mL
Description
SLC14A1 Monoclonal antibody specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC14A1
Monoclonal
DyLight 550
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
6563
Store at 4°C in the dark.
IgG2b
Flow Cytometry, CyTOF
888418
Flow Cytometry, CyTOF-ready
FLJ33745, HsT1341, HUT11, JKFLJ41687, RACH1, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1UT-B1, UTEblood group Kidd urea transporter
NS0 mouse myeloma cell line transfected with human SLC14A1, Accession # Q13336
0.1 mL