Learn More
SLC16A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NBP1-81251
 View more versions of this product
Product Code. 18707913
  • € 572.00 / 0.10mL
Description
SLC16A3 Polyclonal specifically detects SLC16A3 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
SLC16A3
RUO
IgG
MCT 3, MCT3MCT 4, MCT4, MCT4MGC138472, MGC138474, Monocarboxylate transporter 3, monocarboxylate transporter 4, solute carrier family 16 (monocarboxylic acid transporters), member 3, Solute carrier family 16 member 3, solute carrier family 16, member 3 (monocarboxylic acid transporter 4)
This antibody was developed against Recombinant Protein corresponding to amino acids:AEEEKLHKPPADSGVDLREVEHFLKAEPEKNGEVVHTPETSV
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.