missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC16A3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 236.00 - € 541.00
Specifications
| Antigen | SLC16A3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231388
|
Novus Biologicals
NBP3-33421-20ul |
20 μL |
€ 236.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230859
|
Novus Biologicals
NBP3-33421-100ul |
100 μL |
€ 541.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC16A3 Monoclonal antibody specifically detects SLC16A3 in Human samples. It is validated for ELISA,Western BlotSpecifications
| SLC16A3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| MCT 3, MCT3MCT 4, MCT4, MCT4MGC138472, MGC138474, Monocarboxylate transporter 3, monocarboxylate transporter 4, solute carrier family 16 (monocarboxylic acid transporters), member 3, Solute carrier family 16 member 3, solute carrier family 16, member 3 (monocarboxylic acid transporter 4) | |
| Recombinant protein of human SLC16A3/MCT4 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 9123 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title