Learn More
Invitrogen™ SLC16A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576688
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
Specifications
| SLC16A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A4 | |
| AW146050; MCT 4; MCT 5; MCT4; MCT5; monocarboxylate transporter; monocarboxylate transporter 4; Monocarboxylate transporter 5; Slc16a4; solute carrier family 16 (monocarboxylic acid transporters), member 4; solute carrier family 16 member 4; solute carrier family 16, member 4; solute carrier family 16, member 4 (monocarboxylic acid transporter 5) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 229699, 9122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15374, Q8R0M8 | |
| SLC16A4 | |
| Recombinant full length Human SLC16A4. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.