missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total SLC16A8.
SLC16A8 is proton-linked monocarboxylate transporter. It catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
Specifications
Specifications
| Antigen | SLC16A8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | SLC16A8 |
| Gene Accession No. | O35308, O70461, O95907 |
| Gene Alias | LOW QUALITY PROTEIN: monocarboxylate transporter 3; MCT 3; MCT3; monocarboxylate transporter; monocarboxylate transporter 3; monocarboxylate transporter 3; LOW QUALITY PROTEIN: monocarboxylate transporter 3; preferentially expressed in RPE cells and choroid plexus epithelium; proton-coupled monocarboxylate transporter 3; REMP; Slc16a8; solute carrier 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 (monocarboxylate transporter), member 8; solute carrier family 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 member 8; solute carrier family 16, member 8 (monocarboxylic acid transporter 3) |
| Gene Symbols | SLC16A8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
