missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC17A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143477
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
SLC17A3 may be involved in actively transporting phosphate into cells via Na(+) cotransport.
Specifications
| SLC17A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A3 | |
| GOUT4; Na(+)/PI cotransporter 4; NPT4; SLC17A3; sodium/phosphate cotransporter 4; Sodium-dependent phosphate transport protein 4; solute carrier family 17 (organic anion transporter), member 3; solute carrier family 17 (sodium phosphate), member 3; solute carrier family 17 member 3; UAQTL4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105355, 10786, 266730 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.92 mg/mL | |
| PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
| O00476 | |
| SLC17A3 | |
| Recombinant protein encompassing a sequence within the center region of human SLC17A3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction