missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC17A4, Mouse anti-Human, Clone: 3E4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010050-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: NLSIAIPAMVNNTAPPSQPNASTERPSTDSQGYWNETLKEFKAMAPAYDWSPEIQGIILSSLNYGSFLAPIPSGYVSpecifications
| SLC17A4 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 17 (sodium phosphate), member 4 | |
| KAIA2138/KIAA2138/MGC129623 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E4 | |
| In 1x PBS, pH 7.4 | |
| NM_005495 | |
| SLC17A4 | |
| SLC17A4 (NP_005486.1, 56 a.a. ∼ 131 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10050 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction