missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC17A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542449
This item is not returnable.
View return policy
Description
Peptide sequence: YFCEYWLFYT IMAYTPTYIS SVLQANLRDS GILSALPFVV GCICIILGGL Sequence homology: Cow: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
As a Na/PO4 cotransporter, SLC17A4 may be important to the regulation of Li transport and its therapeutic effects.
Specifications
| SLC17A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC17A4 | |
| 9130214H05Rik; KAIA2138; Na/PO4 cotransporter; probable small intestine urate exporter; putative small intestine sodium-dependent phosphate transport protein; Slc17a4; solute carrier family 17 (sodium phosphate), member 4; solute carrier family 17 member 4; solute carrier family 17, member 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10050 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y2C5 | |
| SLC17A4 | |
| Synthetic peptide directed towards the middle region of human SLC17A4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction