missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC17A5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13415017
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13415017 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13415017 Supplier Invitrogen™ Supplier No. PA530517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat. Predicted reactivity: Mouse (88%), Rat (89%), Rabbit (89%), Chicken (81%), Sheep (86%), Rhesus Monkey (99%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC17A5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SLC17A5
Gene Accession No. Q8BN82, Q9NRA2
Gene Alias 4631416G20Rik; 4732491M05; AST; FLJ22227; FLJ23268; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; hp59; ISSD; membrane glycoprotein HP59; Membrane glycoprotein SP55; NSD; proton-coupled sialic acid transporter; sb:cb809; SD; sialic acid storage disease; sialin; SIASD; SLC17A5; SLD; Sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; solute carrier family 17 (anion/sugar transporter), member 5; solute carrier family 17 member 5; SP55; Vesicular H(+)/Aspartate-glutamate cotransporter; zgc:153077
Gene Symbols SLC17A5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC17A5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235504, 26503
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.