missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Involved in vesicular storage and exocytosis of ATP. May accumulate ATP and other nucleotides in secretory vesicles such as adrenal chromaffin granules and synaptic vesicles.
Specifications
Specifications
| Antigen | SLC17A9 |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SLC17A9 |
| Gene Accession No. | Q8VCL5, Q9BYT1 |
| Gene Alias | C20orf59, SLC17A9, Vesicular nucleotide transporter, VNUT |
| Gene Symbols | SLC17A9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?