missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC18A2 Mouse anti-Human, Clone: OTI10C11, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500463S
This item is not returnable.
View return policy
Description
SLC18A2 Monoclonal antibody specifically detects SLC18A2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SLC18A2 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q05940 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6571 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10C11 | |
| Unconjugated | |
| SLC18A2 | |
| 1110037L13Rik; 9330105E13; MGC120477; MGC120478; MGC26538; MNAT; monoamine neurotransmitter transporter; monoamine transporter; SLC18A2; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); solute carrier family 18 member 2; solute carrier family 18 member A2; solute carrier family 18, member 2; Svat; SVMT; synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; vesicular amine transporter 2; vesicular monoamine transporter 2; VMAT2; VMAT-2 | |
| Full-length protein expressed in 293T cell transfected with human SLC18A2 expression vector | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction