missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC18A2 Mouse anti-Human, Clone: OTI10C11, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500463
This item is not returnable.
View return policy
Description
VMAT2 is involved in the ATP-dependent vesicular transport of biogenic amine neurotransmitters. pumps cytosolic monoamines including dopamine, norepinephrine, serotonin, and histamine into synaptic vesicles. It is a requisite for vesicular amine storage prior to secretion via exocytosis.Specifications
| SLC18A2 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q05940 | |
| SLC18A2 | |
| Full-length protein expressed in 293T cell transfected with human SLC18A2 expression vector | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10C11 | |
| Unconjugated | |
| SLC18A2 | |
| SVAT, SVMT, VAT2, VMAT2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6571 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction