missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A1 Mouse anti-Human, Rat, Biotin, Clone:S180-41, Abnova™
Mouse monoclonal antibody raised against full length recombinant human SLC1A1.
Brand: Abnova MAB17014.100ug
This item is not returnable.
View return policy
Specifications
| SLC1A1 | |
| Monoclonal | |
| Biotin | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| SLC1A1 | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| S180-41 | |
| solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| EAAC1, EAAT3 | |
| Recombinant protein corresponding to full length human SLC1A1. | |
| 6505 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction