missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A5 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32239AF405
This item is not returnable.
View return policy
Description
SLC1A5 Polyclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Western Blot
Specifications
| SLC1A5 | |
| Polyclonal | |
| Western Blot | |
| AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
| The immunogen this antibody was made to, maps to a region between residue 450 to 500 of human Sodium-Dependent Neutral Amino Acid Transporter Type 2 using the numbering given in entry NP_005619.1 (GeneID 6510). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6510 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction