missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC20A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12423-1-AP-150UL
This item is not returnable.
View return policy
Description
Retrovirus receptors allow infection of human and murine cells by various retroviruses. The receptors that have been identified at the molecular level include CD4 for human immunodeficiency virus, Rec1 for murine ecotropic virus, and GLVR1 for gibbon ape leukemia virus. These 3 proteins show no homology to one another at the DNA or protein level. GLVR1 is a sodium-dependent phosphate symporter.Specifications
| SLC20A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC20A1 | |
| Glvr 1, GLVR1, Phosphate transporter 1, PiT 1, PIT1, SLC20A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20515, 6574, 81826 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q61609, Q8WUM9, Q9JJP0 | |
| SLC20A1 | |
| SLC20A1 Fusion Protein Ag3084 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction