missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A1 Antibody (609222), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6469M
This item is not returnable.
View return policy
Description
SLC22A1 Monoclonal antibody specifically detects SLC22A1 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| SLC22A1 | |
| Monoclonal | |
| DyLight 594 | |
| hOCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1 | |
| NS0 mouse myeloma cell line transfected with human SLC22A1, Met1-Thr554, Accession # NP_003048 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 609222 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6580 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction