missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A1 Antibody (SR2179), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | SLC22A1 |
|---|---|
| Clone | SR2179 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:30 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18689774
|
Novus Biologicals
NBP3-22126-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673434
|
Novus Biologicals
NBP3-22126-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
SLC22A1 Monoclonal antibody specifically detects SLC22A1 in Human samples. It is validated for Western Blot, Flow CytometrySpezifikation
| SLC22A1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6580 | |
| IgG | |
| Affinity purified |
| SR2179 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| hOCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1 | |
| A synthesized peptide derived from human SLC22A1 (Uniprot #: O15245) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts