missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24617-1-AP-20UL
This item is not returnable.
View return policy
Description
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.Specifications
| SLC22A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A1 | |
| HOCT1, OCT1, oct1_cds, Organic cation transporter 1, SLC22A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20517, 24904, 6580 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08966, O15245, Q63089 | |
| Slc22a1 | |
| SLC22A1 Fusion Protein Ag18806 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction