missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569255
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
SLC22A10 gene ontology annotations related to this gene include transmembrane transporter activity.
Specifications
| SLC22A10 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A10 | |
| hOAT5; OAT5; Organic anion transporter 5; SLC22A10; solute carrier family 22 (organic anion/cation transporter), member 10; solute carrier family 22 member 10; solute carrier family 22, member 10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 387775 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q63ZE4 | |
| SLC22A10 | |
| synthetic peptide directed towards the middle region of human SLC22A10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?