missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A12 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97265AF405
This item is not returnable.
View return policy
Description
SLC22A12 Polyclonal antibody specifically detects SLC22A12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC22A12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 116085 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| OATL4, Organic anion transporter 4-like protein, Renal-specific transporter, RSTmember 12, solute carrier family 22 (organic anion/urate transporter), member 12, URAT1, URAT1Urate anion exchanger 1, urate transporter 1 | |
| Produced in rabbits immunized with E. coli-derived Human SLC22A12 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto