missing translation for 'onlineSavingsMsg'
Learn More

SLC22A16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 292.00 - € 572.00

Specifications

Antigen SLC22A16
Concentration 0.05mg/mL
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18406171
View Documents
Novus Biologicals
NBP1-85410-25ul
25 μL
€ 292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18279846
View Documents
Novus Biologicals
NBP1-85410
0.1 mL
€ 572.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC22A16 Polyclonal specifically detects SLC22A16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SLC22A16
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Human
Carnitine transporter 2, CT2Organic cation/carnitine transporter 6, Flipt 2, FLIPT2member 16, fly-like putative organic ion transporter 2, Fly-like putative transporter 2, organic cation transporter 6, Organic cation transporter OKB1, solute carrier family 22 (organic cation/carnitine transporter), member 16, solute carrier family 22 member 16
SLC22A16
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
0.05mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
85413
This antibody was developed against Recombinant Protein corresponding to amino acids:KLPETLGKRLATTWEEAAKLESENESKSSKLLLTTNNSGLEKTEAITPRDSGLGE
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.