missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SLC22A16 (Human) Recombinant Protein

Product Code. 16146323
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
This item is not returnable. View return policy

Product Code. 16146323

Brand: Abnova™ H00085413G01.2ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human SLC22A16 full-length ORF (ADZ15999.1) recombinant protein without tag.

  • Sequence: MGSRHFEGIYDHVGHFGRFQRVLYFICAFQNISCGIHYLASVFMGVTPHHVCRPPGNVSQVVFHNHSNWSLEDTGALLSSGQKDYVTVQLQNGEIWELSRCSRNKRENTSSLGYEYTGSKKEFPCVDGYIYDQNTWKSTAVTQWNLVCDRKWLAMLIQPLFMFGVLLGSVTFGYFSDRLGRRVVLWATSSSMFLFGIAAAFAVDYYTFMAARFFLAMVASGYLVVGFVYVMEFIGMKSRTWASVHLHSFFAVGTLLVALTGYLVRTWWLYQMILSTVTVPFILCCWVLPETPFWLLSEGRYEEAQKIVDIMAKWNRASSCKLSELLSLDLQGPVSNSPTEVQKHNLSYLFYNWSITKRTLTVWLIWFTGSLGFYSFSLNSVNLGGNEYLNLFLLGVVEIPAYTFVCIAMDKVGRRTVLAYSLFCSALACGVVMVIPQKHYILGVVTAMVGKFAIGAAFGLIYLYTAELYPTIVRSLAVGSGSMVCRLASILAPFSVDLSSIWIFIPQLFVGTMALLSGVLTLKLPETLGKRLATTWEEAAKLESENESKSSKLLLTTNNSGLEKTEAITPRDSGLGE

Specifications

Accession Number ADZ15999.1
Gene ID (Entrez) 85413
Name solute carrier family 22 (organic cation/carnitine transporter), member 16
Preparation Method Wheat germ expression system
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CT2, FLIPT2, OCT6, OKB1, dJ261K5.1
Gene Symbol SLC22A16
Species Wheat Germ (in vitro)
Buffer 25 mM Tris-HCl of pH 8.0 containing 2% glycerol
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.