missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A16 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142244
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes a member of the organic zwitterion transporter protein family which transports carnitine. The encoded protein has also been shown to transport anticancer drugs like bleomycin successful treatment has been correlated with the level of activity of this transporter in tumor cells.
Specifications
| SLC22A16 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A16 | |
| Carnitine transporter 2; CT2; dJ261K5.1; epididymis secretory protein Li 18; flipt 2; FLIPT2; fly-like putative organic ion transporter 2; fly-like putative transporter 2; HEL-S-18; OAT6; OCT6; OKB1; organic cation transporter 6; organic cation transporter OKB1; Organic cation/carnitine transporter 6; SLC22A16; solute carrier family 22 (organic cation transporter), member 16; solute carrier family 22 (organic cation/carnitine transporter), member 16; solute carrier family 22 member 16; WUGSC:RG331P03.1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 85413 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q86VW1 | |
| SLC22A16 | |
| Peptide with sequence CSRNKRENTSSL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction