missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A18, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00005002-B01P.50ug
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Sequence: MQGARAPRDQGQSPGRMSALGRSSVILLTYVLAATELTCLFMQFSIVPYLSRKLGLDSIAFGYLQTTFGVLQLLGGPVFGRFADQRGARAALTLSFLAALALYLLLAAASSPALPGVYLLFASRLPGALMHTLPAAQMVITDLSAPEERPAALGRLGLCFGVGVILGSLLGGTLVSAYGIQCPAILAALATLLGAVLSFTCIPASTKGAKTDAQAPLPGGPRASVFDLKAIASLLRLPDVPRIFLVKVASNCPTGLFMVMFSIISMDFFQLEAAQAGYLMSFFGLLQMVTQGLVIGQLSSHFSEEVLLRASVLVFIVVGLAMAWMSSVFHFCLLVPGLVFSLCTLNVVTDSMLIKAVSTSDTGTMLGLCASVQPLLRTLGPTVGGLLYRSFGVPVFGHVQVAINTLVLLVLWRKPMPQRKDKVRTekniske data
| SLC22A18 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_002555.3 | |
| SLC22A18 | |
| SLC22A18 (NP_002546.2, 1 a.a. ∼ 424 a.a) full-length human protein. | |
| RUO | |
| 5002 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| solute carrier family 22, member 18 | |
| BWR1A/BWSCR1A/DKFZp667A184/HET/IMPT1/ITM/ORCTL2/SLC22A1L/TSSC5/p45-BWR1A | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion