missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A3 Recombinant Rabbit Monoclonal Antibody (JE53-70)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536158
This item is not returnable.
View return policy
Description
SLC22A3 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq, Jul 2008].
Specifications
| SLC22A3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75751 | |
| SLC22A3 | |
| Recombinant protein within Human SLC22A3 aa 281-470. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE53-70 | |
| Unconjugated | |
| SLC22A3 | |
| 3-Oct; EMT; EMT organic cation transporter 3; EMTH; extraneuronal monoamine extraneuronal monoamine; extraneuronal monoamine transporter; Oct3; Orct3; Organic cation transporter 3; Slc22a3; Slca22a3; solute carrier family 22 (extraneuronal monoamine transporter), member 3; solute carrier family 22 (organic cation transporter), member 3; solute carrier family 22 member 3; solute carrier family 22, member 3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction