missing translation for 'onlineSavingsMsg'
Learn More

SLC22A8 Antibody - BSA Free, Novus Biologicals™

Product Code. 18617310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18617310 0.1 mL 0.1mL
18666891 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18617310 Supplier Novus Biologicals Supplier No. NBP2946040.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC22A8 Polyclonal antibody specifically detects SLC22A8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias hOAT3, MGC24086, OAT3Organic anion transporter 3, solute carrier family 22 (organic anion transporter), member 8, solute carrier family 22 member 8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 443-542 of human SLC22A8 (NP_004245.2). QTGMGVSNLWTRVGSMVSPLVKITGEVQPFIPNIIYGITALLGGSAALFLPETLNQPLPETIEDLENWSLRAKKPKQEPEVEKASQRIPLQPHGPGLGSS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9376
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.