missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC23A2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17131214 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17131214 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17131214 Supplier Invitrogen™ Supplier No. PA5113352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC23A2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1.The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC23A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC23A2
Gene Accession No. Q9UGH3
Gene Alias AI844736; hSVCT2; Kiaa0238; LOW QUALITY PROTEIN: solute carrier family 23 member 2; membrane transporter; mKIAA0238; mSVCT2; Na(+)/L-ascorbic acid transporter 2; NBTL1; Nucleobase transporter-like 1 protein; Slc23a1; SLC23A2; sodium-coupled ascorbic acid transporter 2; sodium-coupled ascorbic acid transporter SVCT2; sodium-dependent vitamin C transporter 2; sodium-dependent vitamin C transporter-2; solute carrier family 23 (ascorbic acid transporter), member 2; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; solute carrier family 23 member 2; SVCT2; SVCT-2; SVCT2-like protein; testicular secretory protein Li 48; yolk sac permease-like molecule 2; YSPL2; YSPL3
Gene Symbols SLC23A2
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 23 member 2 protein (1-90AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9962
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.