missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC23A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95153-0.1ml
This item is not returnable.
View return policy
Description
SLC23A3 Polyclonal antibody specifically detects SLC23A3 in Mouse samples. It is validated for Western Blot
Specifications
| SLC23A3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| E2-binding protein 3, E2BP3, FLJ31168, Na(+)/L-ascorbic acid transporter 3, Sodium-dependent vitamin C transporter 3, solute carrier family 23 (nucleobase transporters), member 3, solute carrier family 23 member 3, SVCT3HPC E2-binding protein 3, Yspl1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 394-493 of human SLC23A3 (NP_653313.3). IPGTQLERGLGQGLPSPFTAQEARMPQKPREKAAQVYRLPFPIQNLCPCIPQPLHCLCPLPEDPGDEEGGSSEPEEMADLLPGSGEPCPESSREGFRSQK | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 151295 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction