missing translation for 'onlineSavingsMsg'
Learn More

SLC24A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15932065
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15932065 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15932065 Supplier Invitrogen Supplier No. PA575537

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SLC24A1 Polyclonal Antibody

SLC24A1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC24A1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC24A1
Gene Accession No. O60721, Q9QZM6
Gene Alias CSNB1D; HsT17412; KIAA0702; Na(+)/K(+)/Ca(2+)-exchange protein 1; NCKX; Nckx1; retinal Na/Ca,K exchanger; retinal rod Na+/Ca+/K+ exchanger; retinal rod Na-Ca+K exchanger; RODX; SLC24A1; sodium/calcium/potassium exchanger; sodium/potassium/calcium exchanger 1; solute carrier family 24 (sodium/potassium/calcium exchanger), member 1; solute carrier family 24 member 1
Gene Symbols SLC24A1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 198-243 of Human SLC24A1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214111, 56814, 9187
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.