missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR4 Antibody (OTI4G1), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74865JF549
This item is not returnable.
View return policy
Description
WDR4 Monoclonal specifically detects WDR4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| WDR4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WDR4 | |
| Full length recombinant protein of human WDR4 (NP_387510) produced in HEK293T cells. | |
| 0.1 mL | |
| 10785.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4G1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| tRNA (guanine-N(7)-)-methyltransferase subunit WDR4, WD repeat domain 4, 10TRM82, WD repeat-containing protein 4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction