missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC24A4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 470.00
Specifications
| Antigen | SLC24A4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621681
|
Novus Biologicals
NBP2-93198-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657490
|
Novus Biologicals
NBP2-93198-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC24A4 Polyclonal antibody specifically detects SLC24A4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SLC24A4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Na(+)/K(+)/Ca(2+)-exchange protein 4, NCKX4FLJ38852, SHEP6, SLC24A2, sodium/potassium/calcium exchanger 4, solute carrier family 24 (sodium/potassium/calcium exchanger), member 4, Solute carrier family 24 member 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human SLC24A4 (NP_705932.2). WWAVCRDSVYYTISVIVLIVFIYDEQIVWWEGLVLIILYVFYILIMKYNVKMQAFFTVKQKSIANGNPVNSELEAGNDFYDGSYDDPSVPLLGQVKEKPQY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 123041 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title