Learn More
SLC25A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15235-1-AP-150UL
 View more versions of this product
Product Code. 16864324
  • € 410.00 / 150µL
Description
This antibody specifically recognized the endogenous SLC25A1; no protein was detected in cells from subjects containing two alleles with truncating mutations with this antibody.
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
Specifications
Specifications
SLC25A1
Polyclonal
Unconjugated
SLC25A1
CIC, Citrate transport protein, CTP, SLC20A3, SLC25A1, Tricarboxylate carrier protein
Rabbit
Antigen Affinity Chromatography
RUO
13358, 29743, 6576
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P32089, P53007, Q8JZU2
SLC25A1
SLC25A1 Fusion Protein Ag7352
150 μL
Primary
Human, Mouse, Rat