missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A12 Antibody (10B12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26321-50ul
This item is not returnable.
View return policy
Description
SLC25A12 Monoclonal antibody specifically detects SLC25A12 in Human samples. It is validated for Western Blot, ELISA
Specifications
| SLC25A12 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8604 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 10B12 | |
| Western Blot 1:500-1:2000, ELISA | |
| AGC1, araceli hiperlarga, ARALAR, ARALAR1, calcium binding mitochondrial carrier superfamily member Aralar1, calcium-binding mitochondrial carrier protein Aralar1, Mitochondrial aspartate glutamate carrier 1, solute carrier family 25 (mitochondrial carrier, Aralar), member 12, Solute carrier family 25 member 12 | |
| A synthesized peptide derived from Human SLC25A12 [UniProt O75746] | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction