missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A14, Mouse anti-Human, Clone: 1C12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009016-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MGIFPGIILIFLRVKFATAAVIVSGHQKSTTVSHEMSGLNWKPFVYGGLASIVAEFGTFPVDLTKTRLQVQGQSIDARFKEIKYRGMFHALFRICKEEGVLALYSGIAPALLRQASYGTIKIGIYQSLKRLFVERLEDETLLINMICGVVSGVISSTIANPTDVLKIRMQAQGSLFQGSMIGSFIDIYQQEGTRGLWRGVVPTAQRAAIVVGVELPVYDITKKHLILSGMMGDTILTHFVSSFTCGLAGALASNPVDVVRTRMMNQRAIVGHVDLYKGTVDGILKMWKHEGFFALYKGFWPNWLRLGPWNIIFFITYEQLKRLQISpecifications
| SLC25A14 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 25 (mitochondrial carrier, brain), member 14 | |
| BMCP1/MGC149543/UCP5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1C12 | |
| In 1x PBS, pH 7.4 | |
| NM_003951.2 | |
| SLC25A14 | |
| SLC25A14 (NP_003942.1, 1 a.a. ∼ 325 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9016 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction