missing translation for 'onlineSavingsMsg'
Learn More

SLC25A20 Antibody, Novus Biologicals™

Product Code. 30229796 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30229796 100 μL 100µL
30230518 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30229796 Supplier Novus Biologicals Supplier No. NBP333453100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SLC25A20 Monoclonal antibody specifically detects SLC25A20 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A20
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Formulation PBS (pH 7.3), 50% glycerol, 0.05% BSA
Gene Alias CACCarnitine/acylcarnitine translocase, CACTSolute carrier family 25 member 20, mitochondrial carnitine/acylcarnitine carrier protein, solute carrier family 25 (carnitine/acylcarnitine translocase), member 20
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC25A20 (NP_000378.1).,, Sequence:, MADQPKPISPLKNLLAGGFGGVCLVFVGHPLDTVKVRLQTQPPSLPGQPPMYSGTFDCFRKTLFREGITGLYRGMAAPIIGVTPMFAVCFFGFGLGKKLQ
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 788
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.