Learn More
SLC25A22 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-25402-100UL
 View more versions of this product
Product Code. 18029895
  • € 450.00 / 100µL
Description
The SLC25A22 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC25A22. This antibody recognizes human, mouse, rat antigen. The SLC25A22 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
SLC25A22
1000 μg/mL
FLJ13044, GC 1, GC1, Glutamate/H(+) symporter 1, SLC25A22
100 μL
79751
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa