missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC25A24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17138883
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17138883 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17138883 Supplier Invitrogen™ Supplier No. PA5110959

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SLWRGNGTNV IKIAPETAVK FWAYEQYKKL LTEEGQKIGT FERFISGSMA GATAQTFIYP MEVMKTRLAV GKTGQYSGIY DCAK Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 95%.

SLC25A24 is a calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. It may act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A24
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A24
Gene Accession No. Q6NUK1
Gene Alias 2610016M12Rik; APC1; Calcium-binding mitochondrial carrier protein SCaMC-1; calcium-binding mitochondrial carrier protein SCaMC-1-like; Calcium-binding mitochondrial carrier protein SCaMC-1-like protein; calcium-binding transporter; DKFZp586G0123; EFINAL; MCSC1; Mitochondrial ATP-Mg/Pi carrier protein 1; mitochondrial Ca(2+)-dependent solute carrier protein 1; Peroxisomal Ca(2+)-dependent solute carrier; peroxisomal Ca-dependent solute carrier; RP11-356N1.3; scamc1; SCAMC-1; short calcium-binding mitochondrial carrier 1; si:ch211-63o20.9; similar to mitochondrial solute carrier; Slc25a24; Small calcium-binding mitochondrial carrier protein 1; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 24; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24; solute carrier family 25 member 24; zgc:92470
Gene Symbols SLC25A24
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SLC25A24. Recombinant protein control fragment (Product #RP-94040).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.