Learn More
SLC25A24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14669-1-AP-20UL
 View more versions of this product
Product Code. 16843424
  • € 145.00 / 20µL
Description
SLC25A24 is a calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. It may act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
Specifications
Specifications
SLC25A24
Polyclonal
Unconjugated
SLC25A24
APC1, MCSC1, SCAMC 1, SCAMC1, SLC25A24
Rabbit
Antigen Affinity Chromatography
RUO
229731, 29957, 310791
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6NUK1, Q8BMD8
SLC25A24
SLC25A24 Fusion Protein Ag6334
20 μL
Primary
Human, Rat, Mouse