missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC25A3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15889763
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15889763 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889763 Supplier Invitrogen™ Supplier No. PA571358

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC25A3 Polyclonal Antibody for Western Blot

The protein encoded by this gene catalyzes the transport of phosphate into the mitochondrial matrix, either by proton cotransport or in exchange for hydroxyl ions. The protein contains three related segments arranged in tandem which are related to those found in other characterized members of the mitochondrial carrier family. Both the N-terminal and C-terminal regions of this protein protrude toward the cytosol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC25A3
Gene Accession No. Q00325
Gene Alias 5730556H19Rik; OK/SW-cl.48; PHC; Phosphate carrier protein, mitochondrial; phosphate carrier, mitochondrial; phosphate transport protein; PTP; Slc25a3; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 3; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 3; solute carrier family 25 member 3
Gene Symbols SLC25A3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SLC25A3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5250
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.