missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106594
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC25A31.
Mitochondrial ADP/ATP carriers, such as SLC25A31, are nuclear-coded mitochondrial proteins that catalyze the exchange of ATP generated in mitochondria by ATP synthase (see MIM 108729) against ADP produced in cytosol by most energy-consuming reactions.
Specifications
| SLC25A31 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A31 | |
| 1700034J06Rik; Aac4; adenine nucleotide translocase 4; adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 4; ADP/ATP translocase 4; ANT 4; Ant4; LOC541168 protein; Sfec; SFEC35kDa; SLC25A31; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 member 31; Sperm flagellar energy carrier protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 689108, 73333, 83447 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3V132, Q9H0C2 | |
| SLC25A31 | |
| A synthesized peptide derived from human SLC25A31(Accession Q9H0C2), corresponding to amino acid residues A135-L185. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction