missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC25A36 Polyclonal Antibody

Product Code. 13406137
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13406137 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13406137 Supplier Invitrogen™ Supplier No. PA531606

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (94%), Xenopus laevis (84%), Pig (95%), Chicken (88%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The SLC25A36 gene is part of the solute carrier family 25 (SLC25), which encodes mitochondrial transport proteins. SLC25A36 specifically functions as a transporter of pyrimidine nucleotides, including deoxynucleotide triphosphates, across the inner mitochondrial membrane. This transport is essential for maintaining mitochondrial DNA synthesis and repair, as well as for the proper functioning of mitochondrial ribonucleotide reductase. The SLC25A36 protein exhibits a characteristic transmembrane structure with six transmembrane helices and a central substrate-binding site. It operates via a mechanism involving substrate binding, conformational changes, and alternating access to the substrate-binding site from either side of the membrane. Dysregulation or mutations in SLC25A36 can lead to mitochondrial dysfunction and are associated with various metabolic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A36
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SLC25A36
Gene Accession No. Q922G0, Q96CQ1
Gene Alias C330005L02Rik; PNC2; Slc25a36; solute carrier family 25 (pyrimidine nucleotide carrier ), member 36; solute carrier family 25 (pyrimidine nucleotide carrier), member 36; solute carrier family 25 member 36; solute carrier family 25, member 36
Gene Symbols SLC25A36
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC25A36. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192287, 55186
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.