missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A41 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98610H
This item is not returnable.
View return policy
Description
SLC25A41 Polyclonal antibody specifically detects SLC25A41 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC25A41 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 284427 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HRP | |
| FLJ40442, MGC34725, solute carrier family 25 member 41, solute carrier family 25, member 41 | |
| Produced in rabbits immunized with E. coli-derived Human SLC25A41 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction