Learn More
SLC25A42 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98611MFV500
Product Code. 18559523
  • € 428.00 / 0.10mL
Description
SLC25A42 Polyclonal antibody specifically detects SLC25A42 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit
Antigen and protein A Affinity-purified
0.1 mL