missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A47 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-57843
This item is not returnable.
View return policy
Description
SLC25A47 Polyclonal specifically detects SLC25A47 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| SLC25A47 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| C14orf68hepatocellular carcinoma-downregulated mitochondrial carrier protein, chromosome 14 open reading frame 68, HDMCPsolute carrier family 25 member 47, Hepatocellular carcinoma down-regulated mitochondrial carrier protein, solute carrier family 25, member 47 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 283600 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 2.5 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_997000.2 | |
| SLC25A47 | |
| Peptide with sequence C-RYGNPDAKPTKAD corresponding to internal region according to NP_997000.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction