missing translation for 'onlineSavingsMsg'
Learn More

SLC25A6 Antibody, Novus Biologicals™

Product Code. 30231754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30231754 20 μL 20µL
30231442 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30231754 Supplier Novus Biologicals Supplier No. NBP33804320ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC25A6 Polyclonal antibody specifically detects SLC25A6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A6
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias Adenine nucleotide translocator 3, ADP, ADP/ATP translocase 3, ADP/ATP translocator of liver, ANT 2, ANT 3, ANT3AAC3, ANT3Y, ATP carrier protein, ATP carrier protein 3, ATP carrier protein, isoform T2, ATP carrier protein, liver, MGC17525, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 6, Solute carrier family 25 member 6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SLC25A6 (NP_001627.2).,, Sequence:, LGGVDKHTQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKSGTEREFRGLGDCLVKITKSDGIRGLYQGFSVSVQGIIIYRAAYFGVYDTAKG
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.