missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51031-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.Specifications
| SLC25A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A6 | |
| AAC3, ADP,ATP carrier protein 3, ADP/ATP translocase 3, ANT 2, ANT 3, ANT3, ANT3Y, SLC25A6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 293, 498885, 67582 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12236, Q5U680 | |
| Slc25a26, Slc25a6 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction