missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC26A6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18056491
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18056491 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18056491 Supplier Invitrogen™ Supplier No. PA5145024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DATANGQEDS KAPDGSTLKA LGLPQPDFHS LILDLGALSF VDTVCLKSLK NIFHDFREIE VEVYMAACHS PVVSQLEAGH FFDASITKKH L Antigen sequence identity: Mouse 33%; Rat 77%.

This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC26A6
Gene Accession No. Q9BXS9
Gene Alias anion exchange transporter; anion transporter 1; B930010B04Rik; CFEX; Chloride-formate exchanger; DKFZp586E1422; Pat1; Pat-1; pendrin L1; pendrin-L1; Pendrin-like protein 1; putative anion transporter-1; SLC26A6; solute carrier family 26 (anion exchanger), member 6; solute carrier family 26 member 6; solute carrier family 26, member 6; sulfate anion transporter
Gene Symbols SLC26A6
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 641-731. Recombinant protein control fragment (Product #RP-109867).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.