missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC26A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121604
This item is not returnable.
View return policy
Description
SLC26A8 Polyclonal Antibody for Western Blot, IHC (P)
SLC26A8 is one member of a family of sulfate/anion transporters. Family members are well conserved in their protein (aa length among species) structures yet have markedly different tissue expression patterns.
Specifications
| SLC26A8 | |
| Polyclonal | |
| Unconjugated | |
| SLC26A8 | |
| Anion exchange transporter; anion transporter/exchanger-8; RP11-482O9.1; SLC26A8; solute carrier family 26 (anion exchanger), member 8; solute carrier family 26 member 8; SPGF3; TAT1; testis anion transporter 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116369 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96RN1 | |
| SLC26A8 | |
| Recombinant protein Testis Anion Transporter 1. The antigen corresponds to amino acid range 664-970 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction