missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC27A1 Monoclonal Antibody (308420)

Product Code. 15553712
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15553712 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15553712 Supplier Invitrogen™ Supplier No. MA524022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody detects human FATP1 in cell transfectants but not non-transfected parent cells in flow cytometry. Reconstitute at 0.5 mg/mL in sterile PBS.

SLC27A1 is a protein coding gene. Mediates the ATP-dependent import of long-chain fatty acids (LCFA) into the cell by mediating their translocation at the plasma membrane. Has also an acyl-CoA ligase activity for long-chain and very-long-chain fatty acids. May act directly as a bona fide transporter, or alternatively, in a cytoplasmic or membrane-associated multimeric protein complex to trap and draw fatty acids towards accumulation. Plays a pivotal role in regulating available LCFA substrates from exogenous sources in tissues undergoing high levels of beta-oxidation or triglyceride synthesis. May be involved in regulation of cholesterol metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 308420
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SLC27A1
Gene Accession No. Q6PCB7
Gene Alias ACSVL5; ACSVL5FATP-1; EC 6.2.1; Fatp; FATP1; FATP-1; Fatty acid transport protein; fatty acid transport protein 1; FLJ00336; long-chain fatty acid transport protein 1; MGC71751; SLC27A1; solute carrier family 27 (fatty acid transporter), member 1; solute carrier family 27 member 1; solute carrier family 27, member 1
Gene Symbols SLC27A1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FATP1 Met1-Leu646.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 376497
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.