missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC27A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14048-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy.Specifications
| SLC27A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC27A2 | |
| ACSVL1, FACVL1, FATP 2, FATP2, Fatty acid transport protein 2, hFACVL1, HsT17226, SLC27A2, THCA CoA ligase, VLACS, VLCS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11001, 26458, 65192 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14975, O35488, P97524 | |
| Slc27a2 | |
| SLC27A2 Fusion Protein Ag5217 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction